missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GUCA1B Partial ORF (NP_002089, 93 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002979-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: KLKWTFKIYDKDGNGCIDRLELLNIVEGIYQLKKACRRELQTEQDQLLTPEEVVDRIFLLVDENGDGQLSLNEFVEGARRDKWVMKMLQMDMNPSSWLAQQRRKSAMFSpecifications
NP_002089 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KLKWTFKIYDKDGNGCIDRLELLNIVEGIYQLKKACRRELQTEQDQLLTPEEVVDRIFLLVDENGDGQLSLNEFVEGARRDKWVMKMLQMDMNPSSWLAQQRRKSAMF | |
RUO | |
GUCA1B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2979 | |
GUCA1B (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E1183/GCAP2/GUCA2 | |
GUCA1B | |
Recombinant | |
wheat germ expression system |