missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GRWD1 Full-length ORF (AAH02440.1, 1 a.a. - 446 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083743-P01.10ug
Additional Details : Weight : 0.00010kg
Description
GRWD1 is a component of the 50S and 80S preribosomal complexes and plays a role in ribosome biogenesis (Gratenstein et al., 2005 [PubMed 15885502]).[supplied by OMIM]
Sequence: MAARKGRRRTCETGEPMEAESGDTSSEGPAQVYLPGRGPPLREGEELVMDEEAYVLYHRAQTGAPCLSFDIVRDHLGDNRTELPLTLYLCAGTQAESAQSNRLMMLRMHNLHGTKPPPSEGSDEEEEEEDEEDEEERKPQLELAMVPHYGGINRVRVSWLGEEPVAGVWSEKGQVEVFALRRLLQVVEEPQALAAFLRDEQAQMKPIFSFAGHMGEGFALDWSPRVTGRLLTGDCQKNIHLWTPTDGGSWHVDQRPFVGHTRSVEDLQWSPTENTVFASCSADASIRIWDIRAAPSKACMLTTATAHDGDVNVISWSRREPFLLSGGDDGALKIWDLRQFKSGSPVATFKQHVAPVTSVEWHPQDSGVFAASGADHQITQWDLAVERDPEAGDVEADPGLADLPQQLLFVHQGETELKELHWHPQCPGLLVSTALSGFTIFRTISVSpecifications
AAH02440.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
75.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GRWD/KIAA1942/RRB1/WDR28 | |
GRWD1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
83743 | |
GRWD1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAARKGRRRTCETGEPMEAESGDTSSEGPAQVYLPGRGPPLREGEELVMDEEAYVLYHRAQTGAPCLSFDIVRDHLGDNRTELPLTLYLCAGTQAESAQSNRLMMLRMHNLHGTKPPPSEGSDEEEEEEDEEDEEERKPQLELAMVPHYGGINRVRVSWLGEEPVAGVWSEKGQVEVFALRRLLQVVEEPQALAAFLRDEQAQMKPIFSFAGHMGEGFALDWSPRVTGRLLTGDCQKNIHLWTPTDGGSWHVDQRPFVGHTRSVEDLQWSPTENTVFASCSADASIRIWDIRAAPSKACMLTTATAHDGDVNVISWSRREPFLLSGGDDGALKIWDLRQFKSGSPVATFKQHVAPVTSVEWHPQDSGVFAASGADHQITQWDLAVERDPEAGDVEADPGLADLPQQLLFVHQGETELKELHWHPQCPGLLVSTALSGFTIFRTISV | |
RUO | |
GRWD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |