missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GPSN2 Full-length ORF (AAH02425.1, 1 a.a. - 308 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | AAH02425.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9524 |
Molecular Weight (g/mol) | 59.62kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173772
|
Abnova™
H00009524-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16183772
|
Abnova™
H00009524-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Microsomal long and very long chain fatty acid elongation uses malonyl-CoA as the 2-carbon donor and consists of 4 sequential reactions. TER catalyzes the final step, reducing trans-2,3-enoyl-CoA to saturated acyl-CoA (Moon and Horton, 2003 [PubMed 12482854]).[supplied by OMIM]
Sequence: MKHYEVEILDAKTREKLCFLDKVEPHATIAEIKNLFTKTHPQWYPARQSLRLDPKGKSLKDEDVLQKLPVGTTATLYFRDLGAQISWVTVFLTEYAGPLFIYLLFYFRVPFIYGHKYDFTSSRHTVVHLACICHSFHYIKRLLETLFVHRFSHGTMPLRNIFKNCTYYWGFAAWMAYYINHPLYTPPTYGAQQVKLALAIFVICQLGNFSIHMALRDLRPAGSKTRKIPYPTKNPFTWLFLLVSCPNYTYEVGSWIGFAIMTQCLPVALFSLVGFTQMTIWAKGKHRSYLKEFRDYPPLRMPIIPFLLSpecifications
AAH02425.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.62kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKHYEVEILDAKTREKLCFLDKVEPHATIAEIKNLFTKTHPQWYPARQSLRLDPKGKSLKDEDVLQKLPVGTTATLYFRDLGAQISWVTVFLTEYAGPLFIYLLFYFRVPFIYGHKYDFTSSRHTVVHLACICHSFHYIKRLLETLFVHRFSHGTMPLRNIFKNCTYYWGFAAWMAYYINHPLYTPPTYGAQQVKLALAIFVICQLGNFSIHMALRDLRPAGSKTRKIPYPTKNPFTWLFLLVSCPNYTYEVGSWIGFAIMTQCLPVALFSLVGFTQMTIWAKGKHRSYLKEFRDYPPLRMPIIPFLL | |
RUO | |
GPSN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9524 | |
GPSN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SC2/TER | |
GPSN2 | |
Recombinant | |
wheat germ expression system |