missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GPR15 Partial ORF (NP_005281.1, 261 a.a. - 360 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002838-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138844

  • 508.00 € / 25µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

Sequence: KFLAIVSGLRQEHYLPSAILQLGMEVSGPLAFANSCVNPFIYYIFDSYIRRAIVHCLCPCLKNYDFGSSTETSDSHLTKALSTFIHAEDFARRRKRSVSL
Specifications

Specifications

NP_005281.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
KFLAIVSGLRQEHYLPSAILQLGMEVSGPLAFANSCVNPFIYYIFDSYIRRAIVHCLCPCLKNYDFGSSTETSDSHLTKALSTFIHAEDFARRRKRSVSL
RUO
GPR15
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2838
GPR15 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC126828/MGC126830
GPR15
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions