Learn More
ACROBiosystems Human GM-CSF R alpha Protein, Fc Tag
Description
GM-CSF R alpha(Granulocyte-macrophage colony-stimulating factor receptor subunit alpha) is also known as CSF2RA, CD116, GMR-alpha. Low affinity receptor for granulocyte-macrophage colony-stimulating factor. Transduces a signal that results in the proliferation, differentiation, and functional activation of hematopoietic cells. Hereditary pulmonary alveolar proteinosis (hPAP) is a rare disorder of pulmonary surfactant accumulation and hypoxemic respiratory failure caused by mutations in CSF2RA (encoding the granulocyte/macrophage colony-stimulating factor [GM-CSF] receptor α-chain [CD116]), which results in reduced GM-CSF-dependent pulmonary surfactant clearance by alveolar macrophages.
Specifications
Specifications
| Accession Number | NP_006131.2 |
| Molecular Weight (g/mol) | 60.9 kDa |
| Quantity | 100 μg |
| Source | Human GM-CSF R alpha, Fc Tag (GRA-H5255) is expressed from human 293 cells (HEK293). It contains AA Glu 23 - Gly 320 (Accession # P15509-1). |
| Immunogen | Glu 23 - Gly 320 |
| Storage Requirements | -20°C |
| Gene Alias | GM-CSF R alpha,GM-CSF-R-alpha,GMCSFR-alpha,GMR-alpha,CSF2RA,CSF2R,CSF2RY,CDw116,CD116 |
| Common Name | GM-CSF R alpha |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.