missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Glucagon R/GCGR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10296-100
This item is not returnable.
View return policy
Description
Glucagon R/GCGR Monoclonal specifically detects Glucagon R/GCGR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.
Specifications
| Glucagon R/GCGR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| FLJ97182, GCGR, GGR, GLR, GL-R, glucagon receptor, GlucagonR, MGC138246 | |
| HEK293 human embryonic kidney cell line transfected with human Glucagon R/GCGR, Met1-Gln142, Accession # P47871 | |
| 100 μg | |
| Primary | |
| Detects human Glucagon R/GCGR in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 591929 | |
| Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 5-25 μg/mL, CyTOF-ready | |
| P47871 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2642 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction