missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GBX2 Partial ORF (NP_001476.2, 141 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002637-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16034055

  • 335.00 € / 10µg
Estimated Shipment: 07-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: AEALQADAEDGKGFLAKEGSLLAFSAAETVQASLVGAVRGQGKDESKVEDDPKGKEESFSLESDVDYSSDDNLTGQAAHKEEDPGHALEE
Specifications

Specifications

NP_001476.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue
AEALQADAEDGKGFLAKEGSLLAFSAAETVQASLVGAVRGQGKDESKVEDDPKGKEESFSLESDVDYSSDDNLTGQAAHKEEDPGHALEE
RUO
GBX2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
2637
GBX2 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GBX2
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions