Learn More
Abnova™ Human GAN Partial ORF (NP_071324, 534 a.a. - 597 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
This gene encodes a member of the cytoskeletal BTB/kelch (Broad-Complex, Tramtrack and Bric a brac) repeat family. The encoded protein plays a role in neurofilament architecture and is involved in mediating the ubiquitination and degradation of some proteins. Defects in this gene are a cause of giant axonal neuropathy (GAN). [provided by RefSeq]
Specifications
Specifications
| Accession Number | NP_071324 |
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| Gene ID (Entrez) | 8139 |
| Molecular Weight (g/mol) | 32.78kDa |
| Name | GAN (Human) Recombinant Protein (Q01) |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 25 μg |
| Immunogen | DLDTGTNYDYVREFKRSTGTWHHTKPLLPSDLRRTGCAALRIANCKLFRLQLQQGLFRIRVHSP |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.