missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human G-CSF Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF-214-NA
This item is not returnable.
View return policy
Description
G-CSF Polyclonal specifically detects G-CSF in Human samples. It is validated for Western Blot, Neutralization.
Specifications
| G-CSF | |
| Polyclonal | |
| Unconjugated | |
| NP_757373 | |
| CSF3 | |
| E. coli-derived recombinant human G-CSF Thr31-Pro204 Accession # NP_757373 | |
| 100 μg | |
| Primary | |
| Detects human G-CSF in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 30% cross-reactivity with recombinant mouse G-CSF and recombinant canine G-CSF is observed. | |
| Human | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 8-48 ng/mL | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3, CSF3OS, Filgrastim, GCSF, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1440 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction