missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FUBP1 Partial ORF (NP_003893, 27 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_003893 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8880 |
Molecular Weight (g/mol) | 37.84kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139545
|
Abnova™
H00008880-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129545
|
Abnova™
H00008880-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase. [provided by RefSeq]
Sequence: VNDAFKDALQRARQIAAKIGGDAGTSLNSNDYGYGGQKRPLEDGDQPDAKKVAPQNDSFGTQLPPMHQQQSRSVMTEEYKVPDGMVGFIIGRGGEQISRIQQESGCKIQISpecifications
NP_003893 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FBP/FUBP | |
FUBP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
8880 | |
FUBP1 (Human) Recombinant Protein (Q01) | |
VNDAFKDALQRARQIAAKIGGDAGTSLNSNDYGYGGQKRPLEDGDQPDAKKVAPQNDSFGTQLPPMHQQQSRSVMTEEYKVPDGMVGFIIGRGGEQISRIQQESGCKIQI | |
RUO | |
FUBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |