Learn More
Human FPRL2/FPR3 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB3896
 View more versions of this product
Product Code. 18101401
  • 437.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
FPRL2/FPR3 Monoclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FPRL2/FPR3
Monoclonal
LYOPH
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
P25089
FPR3
HEK293 human embryonic kidney cell line transfected with human FPRL2 Met1-Met352 Accession # P25089
100 μg
Primary
Detects human FPRL2. Stains human FPRL2 transfectants but not irrelevant transfectants.
Human
Purified
Flow Cytometry, CyTOF
374822
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPR3, FPRH1, FPRH1FPRH2, FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2359
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b