missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FOLR3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5319-SP
This item is not returnable.
View return policy
Description
FOLR3 Monoclonal specifically detects FOLR3 in Human samples. It is validated for Western Blot.
Specifications
| FOLR3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_000795 | |
| FOLR3 | |
| Mouse myeloma cell line NS0-derived recombinant human FOLR3 long isoform Ala22-Ser245 Accession # NP_000795 | |
| 25 μg | |
| Primary | |
| Detects human FOLR3 in direct ELISAs and Western blots. In direct ELISAs, 10% cross-reactivity with recombinant human (rh) FOLR2 is observed and no cross-reactivity with rhFOLR4 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 553701 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Folate receptor 3, folate receptor 3 (gamma), folate receptor gamma, FR-G, FR-gamma, gamma-hFR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2352 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction