missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human FLRT2 ELISA Kit
Description
Human FLRT2 quantitates human FLRT2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FLRT2.
This gene encodes a member of the fibronectin leucine rich transmembrane protein (FLRT) family. FLRT family members may function in cell adhesion and/or receptor signalling. Their protein structures resemble small leucine-rich proteoglycans found in the extracellular matrix.
Specifications
Specifications
| Accession Number | Fibronectin leucine rich transmembrane protein 2; fibronectin-like domain-containing leucine-rich transmembrane protein 2; FLRT2; Kiaa0405; Leucine-rich repeat transmembrane protein FLRT2; UNQ232/PRO265 |
| Assay Range | 1.6 to 400 ng/mL |
| Assay Sensitivity | 1.6 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 23768 |
| Gene Symbol | FLRT2 |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?