missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ferritin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB93541-100
This item is not returnable.
View return policy
Description
Ferritin Monoclonal specifically detects Ferritin in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Ferritin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| apoferritin, Cell proliferation-inducing gene 15 protein, Ferritin H subunit, ferritin heavy chain, ferritin, heavy polypeptide 1, FHC, FTHEC 1.16.3.1, FTHL6MGC104426, PIG15, placenta immunoregulatory factor, PLIF, proliferation-inducing protein 15 | |
| Human Ferritin synthetic peptide Accession # P02794 | |
| 100 μg | |
| Primary | |
| Detects human Ferritin in direct ELISAs and Western blots. | |
| Human | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| 962609 | |
| Western Blot 5 ug/mL, Immunocytochemistry 1 ug/mL | |
| P02794 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2495 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction