Learn More
Abnova™ Human FCRL3 Partial ORF (NP_001019838, 625 a.a. - 723 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00115352-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the immunoglobulin receptor superfamily and is one of several Fc receptor-like glycoproteins clustered on the long arm of chromosome 1. The encoded protein contains immunoreceptor-tyrosine activation motifs and immunoreceptor-tyrosine inhibitory motifs in its cytoplasmic domain and may play a role in regulation of the immune system. Mutations in this gene have been associated with rheumatoid arthritis, autoimmune thyroid disease, and systemic lupus erythematosus. [provided by RefSeq]
Sequence: SRPSRIDPQEPTHSKPLAPMELEPMYSNVNPGDSNPIYSQIWSIQHTKENSANCPMMHQEHEELTVLYSELKKTHPDDSAGEASSRGRAHEEDDEENYESpecifications
NP_001019838 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SRPSRIDPQEPTHSKPLAPMELEPMYSNVNPGDSNPIYSQIWSIQHTKENSANCPMMHQEHEELTVLYSELKKTHPDDSAGEASSRGRAHEEDDEENYE | |
RUO | |
FCRL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
115352 | |
FCRL3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FCRH3/IFGP3/IRTA3/SPAP2 | |
FCRL3 | |
Recombinant | |
wheat germ expression system |