missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Fc gamma RIIA (CD32a) (167 Arg/His) His-tag, Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ A42527
Additional Details : Weight : 0.01060kg
Description
This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.Specifications
P12318 | |
PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 | |
Human Fc gamma RIIA (CD32a) (167 Arg/His) His-tag | |
HEK293 cells | |
RUO | |
CD16; CD32; CD32A; CDw32; Fc fragment of IgG low affinity IIa receptor for (CD32); Fc fragment of IgG receptor IIa; Fc fragment of IgG, low affinity IIa, receptor; Fc fragment of IgG, low affinity IIa, receptor (CD32); Fc gamma receptor IIa; Fc receptor, IgG, low affinity III; FCG2; fc-gamma RII-a; Fc-gamma RIII; fc-gamma-RIIa; FcGR; FCGR2; FCGR2A; FCGR2A1; Fcgr3; Fcgr3a; fcRII-a; fcRIII; FGCR; IGFR2; IgG Fc receptor II-a; igG Fc receptor III; Immunoglobulin G Fc receptor II; low affinity immunoglobulin gamma Fc region receptor II-a; Low affinity immunoglobulin gamma Fc region receptor III | |
Human | |
Recombinant | |
≥ 90% |
Control, ELISA, Immunohistochemistry, Western Blot | |
2212 | |
50 μg | |
-20°C | |
<1 EU/μg | |
FCGR2A | |
Unconjugated | |
Lyophilized |