missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Fatty Acid Synthase/FASN Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5927
This item is not returnable.
View return policy
Description
Fatty Acid Synthase/FASN Polyclonal specifically detects Fatty Acid Synthase/FASN in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| Fatty Acid Synthase/FASN | |
| Polyclonal | |
| Unconjugated | |
| P49327 | |
| FASN | |
| E. coli-derived recombinant human Fatty Acid Synthase/FASN Ser10-Cys212 Accession # P49327 | |
| 100 μg | |
| Primary | |
| Detects human Fatty Acid Synthase/FASN in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| EC 2.3.1, EC 2.3.1.85, EC 6.3.3.1, FASN, FASshort chain dehydrogenase/reductase family 27X, member 1, fatty acid synthase, MGC14367, MGC15706, OA-519, SDR27X1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2194 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction