Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2871-500
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Western Blot, Immunoaffinity Purification.
Specifications
| Erythropoietin/EPO | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in PBS. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2056 | |
| Human | |
| Supernatant |
| Western Blot, Affinity Purification | |
| AE7A5 | |
| Western Blot 1 ug/mL, Immunoaffinity Purification | |
| P01588 | |
| EPO | |
| Erythropoietin/EPO synthetic peptide analog, Ala28-Thr53, Accession # P01588 | |
| 500 μg | |
| Primary | |
| Recognizes an epitope within the first 26 amino acids at the NH2 terminus of human urinary and human recombinant EPO. Binds human and rat EPO but does not bind sheep, baboon, or dog EPO (1, 2). | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG2a |