missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Ephrin-A5 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3743-SP
This item is not returnable.
View return policy
Description
Ephrin-A5 Polyclonal specifically detects Ephrin-A5 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Ephrin-A5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AF1, AL-1, EFL-5, EFNA5, EPH-related receptor tyrosine kinase ligand 7, ephrin-A5, EphrinA5, EPLG7, GLC1M, LERK-7, LERK7EFL5, RAGS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1946 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P52803 | |
| EFNA5 | |
| Mouse myeloma cell line NS0-derived recombinant human Ephrin-A5 Gln21-Asn203 Accession # P52803 | |
| 25 μg | |
| Primary | |
| Detects human Ephrin-A5 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) Ephrin-A1, rmEphrin-A2, recombinant human (rh) Ephrin-A3, rhEphrin-A4, rhEphrin-B3, rmEphrin-A4, rmEphrin-B1 and rmEphrin-B2 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction