Learn More
R&D Systems™ Human EMAP-II (NP_001135887.1) VersaClone cDNA
Product Code. 18723267
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18723267 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18723267 Supplier R&D Systems™ Supplier No. RDC3570
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    309.75 € / 10µg
    Save 18.25 €
    5% Off
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
Specifications
Gene EMAP-II/AIMP1
Accession Number NP_001135887.1
Gene ID (Entrez) 9255
Gene Alias AIMP1, aminoacyl tRNA synthetase complex-interacting multifunctional protein 1, EMAP-2, EMAPII, EMAPIImember 1 (endothelialmonocyte-activating), endothelial-monocyte activating polypeptide II, Multisynthase complex auxiliary component p43, multisynthetase complex auxiliary component p43, SCYE1
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg