missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eIF4ENIF1 (aa 383-517) Control Fragment Recombinant Protein

Product Code. 30207792
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30207792 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30207792 Supplier Invitrogen™ Supplier No. RP91484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51614 (PA5-51614. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a nucleocytoplasmic shuttle protein for the translation initiation factor eIF4E. This shuttle protein interacts with the importin alpha-beta complex to mediate nuclear import of eIF4E. It is predominantly cytoplasmic; its own nuclear import is regulated by a nuclear localization signal and nuclear export signals. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NRA8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 56478
Name Human eIF4ENIF1 (aa 383-517) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610509L04Rik; 4 E-T; A930019J01Rik; AA410001; AU021239; Clast4; D11Ertd166e; eIF4E transporter; EIF4ENIF1; eIF4E-transporter; Eukaryotic translation initiation factor 4 E nuclear import factor 1; eukaryotic translation initiation factor 4 E transporter; Eukaryotic translation initiation factor 4 E transporter-like protein; FLJ21601; FLJ26551; OTTHUMP00000063276; RGD1560908
Common Name eIF4ENIF1
Gene Symbol EIF4ENIF1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FAPIPLEDHAENKVDILEMLQKAKVDLKPLLSSLSANKEKLKESSHSGVVLSVEEVEAGLKGLKVDQQVKNSTPFMAEHLEETLSAVTNNRQLKKDGDMTAFNKLVSTMKASGTLPSQPKVSRNLESHLMSPAEI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.