missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DUSP22 Partial ORF (NP_064570, 112 a.a. - 184 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056940-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: EDALHTVRAGRSCANPNVGFQRQLQEFEKHEVHQYRQWLKEEYGESPLQDAEEAKNILAAPGILKFWAFLRRLSpecifications
NP_064570 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.77kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EDALHTVRAGRSCANPNVGFQRQLQEFEKHEVHQYRQWLKEEYGESPLQDAEEAKNILAAPGILKFWAFLRRL | |
RUO | |
DUSP22 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56940 | |
DUSP22 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
JKAP/JSP1/MKPX/VHX | |
DUSP22 | |
Recombinant | |
wheat germ expression system |