missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DR4 (aa 105-139) Control Fragment Recombinant Protein

Product Code. 30204862
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30204862 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30204862 Supplier Invitrogen™ Supplier No. RP101169

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84201 (PA5-84201. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00220
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8797
Name Human DR4 (aa 105-139) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias APO2; CD261; cytotoxic TRAIL receptor; death receptor 4; DR4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF receptor superfamily member 10 A; TNF-related apoptosis-inducing ligand receptor 1; TNFRSF10A; TRAIL R1; TRAIL receptor 1; TRAILR1; TRAIL-R1; TRAILR-1; tumor necrosis factor receptor superfamily member 10 A; tumor necrosis factor receptor superfamily member 10 A variant 2; tumor necrosis factor receptor superfamily, member 10 A
Common Name DR4
Gene Symbol TNFRSF10A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSSAATIKLHDQSIGTQQWEHSPLGELCPPGSHRS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.