missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human DR3 / TNFRSF25 Protein, Fc Tag

Product Code. 15847817
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847817 1 mg 1mg
15837817 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15847817 Supplier ACROBiosystems Supplier No. TN5H52581MG

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Tumor necrosis factor receptor superfamily member 25 (TNFRSF25) is also known as Apo-3, Death receptor 3 (DDR3 or DR3), Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor TRAMP, Lymphocyte-associated receptor of death, Apo-3, which is a member of the TNF-receptor superfamily. TNFRSF25 is a homodimer protein, which can Interact strongly via the death domains with TNFRSF1 and TRADD to activate at least two distinct signaling cascades, apoptosis and NF-kappa-B signaling. TNFRSF25 is receptor for TNFSF12 / APO3L / TWEAK.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.5 kDa. The protein migrates as 53-62 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number AAI17190
Molecular Weight (g/mol) 45.5 kDa
Quantity 1 mg
Source Human DR3, Fc Tag (TN5-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 199 (Accession # AAI17190).
Immunogen Gln 25 - Gln 199
Storage Requirements -20°C
Gene Alias TNFRSF25,DR3,APO3,DDR3,TNFRSF12,WSL,WSL1
Common Name DR3 / TNFRSF25
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer Tris with Glycine, Arginine and NaCl, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Other Recombinant Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.