missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Dopamine beta-Hydroxylase Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7376
This item is not returnable.
View return policy
Description
Dopamine beta-Hydroxylase Polyclonal specifically detects Dopamine beta-Hydroxylase in Human samples. It is validated for Western Blot.
Specifications
| Dopamine beta-Hydroxylase | |
| Polyclonal | |
| Unconjugated | |
| P09172 | |
| DBH | |
| Chinese hamster ovary cell line CHO-derived recombinant human Dopamine beta-Hydroxylase Leu37-Gly617 Accession # P09172 | |
| 100 μg | |
| Primary | |
| Detects human Dopamine beta-Hydroxylase in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| DBH, DBM, dopamine beta-hydroxylase, Dopamine betaHydroxylase, dopamine beta-hydroxylase (dopamine beta-monooxygenase), Dopamine beta-monooxygenase, DOPBHY, EC 1.14.17.1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 1621 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction