missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human DICER1 Partial ORF (NP_803187, 1813 a.a. - 1912 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_803187 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23405 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16147272
|
Abnova™
H00023405-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16137272
|
Abnova™
H00023405-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 04-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein possessing an RNA helicase motif containing a DEXH box in its amino terminus and an RNA motif in the carboxy terminus. The encoded protein functions as a ribonuclease and is required by the RNA interference and small temporal RNA (stRNA) pathways to produce the active small RNA component that represses gene expression. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq]
Sequence: ESLAGAIYMDSGMSLETVWQVYYPMMRPLIEKFSANVPRSPVRELLEMEPETAKFSPAERTYDGKVRVTVEVVGKGKFKGVGRSYRIAKSAAARRALRSLSpecifications
NP_803187 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DCR1/Dicer/HERNA/KIAA0928 | |
DICER1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
23405 | |
DICER1 (Human) Recombinant Protein (Q01) | |
ESLAGAIYMDSGMSLETVWQVYYPMMRPLIEKFSANVPRSPVRELLEMEPETAKFSPAERTYDGKVRVTVEVVGKGKFKGVGRSYRIAKSAAARRALRSL | |
RUO | |
DICER1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |