missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Assay Range | 20 - 4000 ng/mL |
| Assay Sensitivity | 10 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Plasma, Serum, Tissue Homogenates |
| Specificity | This assay has high sensitivity and excellent specificity for detection of human DHEA-Sulfate. No significant cross-reactivity or interference between human DHEA-Sulfate and analogues was observed. |
| For Use With (Application) | ELISA |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C. |
| Synonym | (3b)-3-(Sulfooxy)androst-5-en-17-one, Androstenolone sulfate, Dehydroepiandrosterone 3b-Sulfate, Dehydroepiandrosterone monosulfate, dehydroepiandrosterone sulfate, Dehydroepiandrosterone sulphate, DHEA Sulfate, DHEA-S, prasterone sulfate, Prasterone-3-sulfate |
| Target | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?