Learn More
Abnova™ Human DDX50 Full-length ORF (NP_076950.1, 1 a.a. - 737 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_076950.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79009 |
Molecular Weight (g/mol) | 109kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184003
|
Abnova™
H00079009-P01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174003
|
Abnova™
H00079009-P01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this DEAD box protein family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box enzyme that may be involved in ribosomal RNA synthesis or processing. This gene and DDX21, also called RH-II/GuA, have similar genomic structures and are in tandem orientation on chromosome 10, suggesting that the two genes arose by gene duplication in evolution. This gene has pseudogenes on chromosomes 2, 3 and 4. Alternative splicing of this gene generates multiple transcript variants, but the full length nature of all the other variants but one has not been defined. [provided by RefSeq]
Sequence: MPGKLLWGDIMELEAPLEESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMKEKLNGDTEEGFNRLSDEFSKSHKSRRKDLPNGDIDEYEKKSKRVSSLDTSTHKSSDNKLEETLTREQKEGAFSNFPISEETIKLLKGRGVTYLFPIQVKTFGPVYEGKDLIAQARTGTGKTFSFAIPLIERLQRNQETIKKSRSPKVLVLAPTRELANQVAKDFKDITRKLSVACFYGGTSYQSQINHIRNGIDILVGTPGRIKDHLQSGRLDLSKLRHVVLDEVDQMLDLGFAEQVEDIIHESYKTDSEDNPQTLLFSATCPQWVYKVAKKYMKSRYEQVDLVGKMTQKAATTVEHLAIQCHWSQRPAVIGDVLQVYSGSEGRAIIFCETKKNVTEMAMNPHIKQNAQCLHGDIAQSQREITLKGFREGSFKVLVATNVAARGLDIPEVDLVIQSSPPQDVESYIHRSGRTGRAGRTGICICFYQPRERGQLRYVEQKAGITFKRVGVPSTMDLVKSKSMDAIRSLASVSYAAVDFFRPSAQRLIEEKGAVDALAAALAHISGASSFEPRSLITSDKGFVTMTLESLEEIQDVSCAWKELNRKLSSNAVSQITRMCLLKGNMGVCFDVPTTESERLQAEWHDSDWILSVPAKLPEIEEYYDGNTSSNSRQRSGWSSGRSGRSGRSGGRSGGRSGRQSRQGSRSGSRQDGRRRSGNRNRSRSGGHKRSFDSpecifications
NP_076950.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
109kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPGKLLWGDIMELEAPLEESESQKKERQKSDRRKSRHHYDSDEKSETRENGVTDDLDAPKAKKSKMKEKLNGDTEEGFNRLSDEFSKSHKSRRKDLPNGDIDEYEKKSKRVSSLDTSTHKSSDNKLEETLTREQKEGAFSNFPISEETIKLLKGRGVTYLFPIQVKTFGPVYEGKDLIAQARTGTGKTFSFAIPLIERLQRNQETIKKSRSPKVLVLAPTRELANQVAKDFKDITRKLSVACFYGGTSYQSQINHIRNGIDILVGTPGRIKDHLQSGRLDLSKLRHVVLDEVDQMLDLGFAEQVEDIIHESYKTDSEDNPQTLLFSATCPQWVYKVAKKYMKSRYEQVDLVGKMTQKAATTVEHLAIQCHWSQRPAVIGDVLQVYSGSEGRAIIFCETKKNVTEMAMNPHIKQNAQCLHGDIAQSQREITLKGFREGSFKVLVATNVAARGLDIPEVDLVIQSSPPQDVESYIHRSGRTGRAGRTGICICFYQPRERGQLRYVEQKAGITFKRVGVPSTMDLVKSKSMDAIRSLASVSYAAVDFFRPSAQRLIEEKGAVDALAAALAHISGASSFEPRSLITSDKGFVTMTLESLEEIQDVSCAWKELNRKLSSNAVSQITRMCLLKGNMGVCFDVPTTESERLQAEWHDSDWILSVPAKLPEIEEYYDGNTSSNSRQRSGWSSGRSGRSGRSGGRSGGRSGRQSRQGSRSGSRQDGRRRSGNRNRSRSGGHKRSFD | |
RUO | |
DDX50 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79009 | |
DDX50 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GU2/GUB/MGC3199/RH-II/GuB | |
DDX50 | |
Recombinant | |
wheat germ expression system |