missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DcR3/TNFRSF6B Antibody, R&D Systems™

Beskrivelse
DcR3/TNFRSF6B Monoclonal specifically detects DcR3/TNFRSF6B in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Tekniske data
Tekniske data
| Antigen | DcR3/TNFRSF6B |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 229817 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | O95407 |
| Gene Alias | DCR3Decoy receptor for Fas ligand, Decoy receptor 3, M68, M68DJ583P15.1.1, TNFRSF6B, TR6, TR6DcR3, tumor necrosis factor receptor superfamily member 6B, tumor necrosis factor receptor superfamily, member 6b, decoy |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?