missing translation for 'onlineSavingsMsg'
Learn More

Human Cystatin B Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18140201
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18140201 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18140201 Supplier R&D Systems Supplier No. MAB1408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin B/Stefin B
Applications Western Blot, Immunoprecipitation, Neutralization, Immunoassay
Classification Monoclonal
Clone 225228
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5.4 ug/mL, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P04080
Gene Alias CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B
Gene Symbols CSTB
Host Species Mouse
Immunogen E. coli-derived recombinant human Cystatin B aa Met2-Phe98 Accession # P04080
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1476
Test Specificity Detects human Cystatin B in Western blots and direct ELISAs. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Cystatins A and S, recombinant mouse Cystatin B, rhFetuin A, and rhHPRG (Histidine-Proline-Rich Glycoprotein) is observed and approximately 5% to 10% cross-reactivity with rhCystatins C, D, E/M, SA, SN, and rhFetuin B is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.