missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CX3CL1 Partial ORF (AAH01163, 26 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006376-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: HHGVTKCNITCSKMTSKIPVALLIHYQQNQASCGKRAIILETRQHRLFCADPKEQWVKDAMQHLDRQAAALTRNGGTFEKQIGEVKPRTTPAAGGMDESVVLEPEATGESSpecifications
AAH01163 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
HHGVTKCNITCSKMTSKIPVALLIHYQQNQASCGKRAIILETRQHRLFCADPKEQWVKDAMQHLDRQAAALTRNGGTFEKQIGEVKPRTTPAAGGMDESVVLEPEATGES | |
RUO | |
CX3CL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
6376 | |
CX3CL1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABCD-3/C3Xkine/CXC3/CXC3C/NTN/NTT/SCYD1/fractalkine/neurotactin | |
CX3CL1 | |
Recombinant | |
wheat germ expression system |