missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CTLA-4 (Research Grade Ipilimumab Biosimilar) PE-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems FAB9928P-100
This item is not returnable.
View return policy
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry.
Specifications
| CTLA-4 | |
| Monoclonal | |
| PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1493 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu111 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
| Human CTLA-4 | |
| 100 Tests | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction