missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CMTM4 Full-length ORF (NP_848933.1, 1 a.a. - 234 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00146223-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138413

  • 335.00 € / 10µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies of signaling molecules. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MRSGEELDGFEGEASSTSMISGASSPYQPTTEPVSQRRGLAGLRCDPDYLRGALGRLKVAQVILALIAFICIETIMACSPCEGLYFFEFVSCSAFVVTGVLLIMFSLNLHMRIPQINWNLTDLVNTGLSAFLFFIASIVLAALNHRAGAEIAAVIFGFLATAAYAVNTFLAVQKWRVSVRQQSTNDYIRARTESRDVDSRPEIQRLDTFSYSTNVTVRKKSPTNLLSLNHWQLA
Specifications

Specifications

NP_848933.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CKLFSF4
CMTM4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
146223
CMTM4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRSGEELDGFEGEASSTSMISGASSPYQPTTEPVSQRRGLAGLRCDPDYLRGALGRLKVAQVILALIAFICIETIMACSPCEGLYFFEFVSCSAFVVTGVLLIMFSLNLHMRIPQINWNLTDLVNTGLSAFLFFIASIVLAALNHRAGAEIAAVIFGFLATAAYAVNTFLAVQKWRVSVRQQSTNDYIRARTESRDVDSRPEIQRLDTFSYSTNVTVRKKSPTNLLSLNHWQLA
RUO
CMTM4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions