missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Claudin-8 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5275
This item is not returnable.
View return policy
Description
Claudin-8 Monoclonal specifically detects Claudin-8 in Human samples. It is validated for Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Claudin-8 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P56748 | |
| CLDN8 | |
| HEK293 human embryonic kidney cell line transfected with human Claudin-8 Met1-Val225 (Ser151Pro) Accession # P56748 | |
| 100 μg | |
| Primary | |
| Detects human Claudin-8. Stains human Claudin-8 transfectants but not irrelevant transfectants. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 474210 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| claudin 8, Claudin8, CLDN8, human CLDN8 gene for claudin-810claudin-8 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9073 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction