missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CIDEC Partial ORF (NP_071377.2, 53 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00063924-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16139486

  • 508.00 € / 25µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DNA fragmentation factor (DFF) induces the fragmentation of DNA associated with apoptosis. A novel family of cell death-inducing DFF45 (MIM 601882)-like effectors (CIDEs), including CIDEC, can also promote apoptosis (Liang et al., 2003 [PubMed 12429024]).[supplied by OMIM]

Sequence: SVRKGIMAYSLEDLLLKVRDTLMLADKPFFLVLEEDGTTVETEEYFQALAGDTVFMVLQKGQKWQPPSEQGTRHPLSLSHKPAKKIDVA
Specifications

Specifications

NP_071377.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.53kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SVRKGIMAYSLEDLLLKVRDTLMLADKPFFLVLEEDGTTVETEEYFQALAGDTVFMVLQKGQKWQPPSEQGTRHPLSLSHKPAKKIDVA
RUO
CIDEC
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
63924
CIDEC (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CIDE-3/FLJ20871/Fsp27
CIDEC
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions