missing translation for 'onlineSavingsMsg'
Learn More

Human CG alpha (HCG alpha) Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18048171
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18048171 0.1 mg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18048171 Supplier R&D Systems Supplier No. R114C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Chorionic Gonadotropin alpha Chain (hCG alpha) Polyclonal specifically detects Chorionic Gonadotropin alpha Chain (hCG alpha) in Human samples. It is validated for .
TRUSTED_SUSTAINABILITY

Specifications

Antigen Chorionic Gonadotropin alpha Chain (HCG alpha)
Classification Polyclonal
Conjugate Unconjugated
Dilution Hidden
Formulation Lyophilized from a 0.2 μm filtered solution in NaH2PO4, NaCl and Sodium Azide with Trehalose. with No Preservative
Gene Alias alpha HCG, CGA, CGA glycoprotein hormones, alpha polypeptide, CG-ALPHA, FSHA, GPHa, GPHa1, HCG, LHA, TSHA
Gene Symbols CGA
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant human Chorionic Gonadotropin, alpha Chain ( alpha HCG)
Purification Method Antigen Affinity-purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1081
Reconstitution Reconstitute with sterile PBS. If 0.5 mL of PBS is used, the antibody concentration will be 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.