Learn More
Invitrogen™ Human CEP63 (aa 378-469) Control Fragment Recombinant Protein
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63530 (PA5-63530. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Accession Number | Q96MT8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 80254 |
| Name | Human CEP63 (aa 378-469) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4921501M07; AL450317.13 gm1; AW107703; CD20R; centrosomal protein 63; centrosomal protein 63 kDa; centrosomal protein of 63 kDa; centrosome protein Cep63; CEP63; D9Mgc41; D9Mgc48e; ET2; LOW QUALITY PROTEIN: centrosomal protein of 63 kDa; SCKL6 |
| Common Name | CEP63 |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.