missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CEP250 (aa 2228-2308) Control Fragment Recombinant Protein

Product Code. 30202296
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202296

Brand: Invitrogen™ RP107382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (60%), Rat (60%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66730 (PA5-66730. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Probably plays an important role in centrosome cohesion during interphase.
TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number Q9BV73
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11190
Name Human CEP250 (aa 2228-2308) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 250 kDa centrosomal protein; AW490617; B230210E21Rik; centrosomal Nek2-associated protein 1; Centrosomal protein 2; centrosomal protein 250; centrosomal protein 250 kDa; centrosome associated protein; centrosome-associated protein CEP250; Cep2; CEP250; CNAP1; C-Nap1; Inmp; Intranuclear matrix protein
Common Name CEP250
Gene Symbol CEP250
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HSPGATSTAELGSRGEQGVQLGEVSGVEAEPSPDGMEKQSWRQRLEHLQQAVARLEIDRSRLQRHNVQLRSTLEQVERERR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.