Learn More
Invitrogen™ Human CEP112 (aa 914-951) Control Fragment Recombinant Protein
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144978 (PA5-144978. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Accession Number | Q8N8E3 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 201134 |
| Name | Human CEP112 (aa 914-951) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1700001M19Rik; 1700029K01Rik; 8430407H02Rik; AV043680; AV207351; CCDC46; centrosomal protein 112; centrosomal protein 112 kDa; centrosomal protein of 112 kDa; centrosomal protein of 112 kDa; LOW QUALITY PROTEIN: centrosomal protein of 112 kDa; Cep112; coiled-coil domain containing 46; coiled-coil domain-containing protein 46; Macoco; RGD1564168 |
| Common Name | CEP112 |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.