missing translation for 'onlineSavingsMsg'
Learn More

Human CEACAM-8/CD66b Antibody, R&D Systems™

Product Code. 18092932
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18092932 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18092932 Supplier R&D Systems Supplier No. AF4246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

CEACAM8/CD66b Polyclonal specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM8/CD66b
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Gene Accession No. P31997
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CEACAM8, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Gene Symbols CEACAM8
Host Species Sheep
Immunogen E. coli-derived recombinant human CEACAM-8/CD66b Gln35-His141 Accession # P31997
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1088
Test Specificity Detects human CEACAM-8/CD66b in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) CEACAM-3, rhCEACAM-6, and rhCEACAM-7 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.