missing translation for 'onlineSavingsMsg'
Learn More

Human CD300b/LMIR5 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18030772
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18030772 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18030772 Supplier R&D Systems Supplier No. AF2879

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

CD300b/LMIR5/CD300LB Polyclonal specifically detects CD300b/LMIR5/CD300LB in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300b/LMIR5/CD300LB
Applications Western Blot, Flow Cytometry, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Gene Accession No. NP_777552
Gene Alias CD300 antigen like family member B, CD300 antigen-like family member B, CD300 molecule-like family member b, CD300B, CD300b antigen, CD300LB, CLM7, CLM-7, CLM7IREM3CD300 molecule like family member b, CMRF35A2, CMRF35-A2, CMRF35-like molecule 7, EC 1.13.11.6, EC 6.3.4.3, Gm253, Immune receptor expressed on myeloid cells 3, IREM-3, Leukocyte mono-Ig-like receptor 5, LMIR5, TREM-5, TREM5CD300b, Triggering receptor expressed on myeloid cells 5
Gene Symbols CD300LB
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CD300b/LMIR5 Ile55-His187 Accession # NP_777552
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124599
Test Specificity Detects human CD300b/LMIR5 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human LMIR2 and recombinant mouse LMIR5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.