Learn More
ACROBiosystems Human CD3 epsilon & CD3 delta Heterodimer Protein, His Tag&Tag Free

Description
T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Specifications
Specifications
| Accession Number | NP_000724.1 (CD3E) and NP_000723.1 (CD3D) |
| Molecular Weight (g/mol) | 16.6 kDa (CD3E) and 13.4 kDa (CD3D) |
| Quantity | 50 μg |
| Source | Human CD3E&CD3D; Heterodimer Protein, His Tag&Tag; Free (CDD-H52W1) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) and P04234-1 (CD3D)). |
| Immunogen | Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) |
| Storage Requirements | -20°C |
| Gene Alias | CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon |
| Common Name | CD3E and CD3D |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.