missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD27 Ligand/TNFSF7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2738-SP
This item is not returnable.
View return policy
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD27 Ligand/TNFSF7/CD70 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| AAA36175 | |
| CD70 | |
| Mouse myeloma cell line NS0-derived recombinant human CD27 Ligand/TNFSF7 Gln45-Pro193 Accession # AAA36175 | |
| 25 μg | |
| Primary | |
| Detects human CD27 Ligand/TNFSF7 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry | |
| 301731 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 970 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction