missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR5 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 11 publications
Brand: R&D Systems MAB181-100
This item is not returnable.
View return policy
Description
CCR5 Monoclonal specifically detects CCR5 in Human samples. It is validated for Immunohistochemistry.
Specifications
| CCR5 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P51681 | |
| CCR5 | |
| NS0 mouse myeloma cell line transfected with human CCR5 Met1-Leu352 Accession # P51681 | |
| 100 μg | |
| Primary | |
| Can be used to detect CCR5 present on stimulated human PBMCs only after cells are fixed with 2% formaldehyde. It does not detect CCR5 on unfixed human PBMCs. It does not cross-react with CCR1, CCR2, or CCR3. For additional information regarding epitope specificity for this antibody and other R&D Systems anti-human CCR5 antibodies, see Lee, B. et al., 1999, J. Biol. Chem. 274:9617. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 45523 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C CKR-5, C-C motif chemokine receptor 5 A159A, CCCKR5, CC-CKR-5FLJ78003, CCR5, CCR-5, CD195, CD195 antigen, chemokine (C-C motif) receptor 5, chemokine receptor CCR5, chemr13, CKR5, CKR-5, HIV-1 fusion coreceptor, IDDM22CMKBR5C-C chemokine receptor type 5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1234 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction