missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL4/MIP-1 beta Quantikine ELISA Kit

Product Code. 18761834
Click to view available options
Quantity:
1 Plate
6 Plates
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18761834

Brand: R&D Systems™ DMB00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human CCL4/MIP-1 beta Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human CCL4/MIP-1 beta Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL4/MIP-1 beta in cell culture supernates (10 ul), serum (150 ul), edta plasma (150 ul), heparin plasma (150 ul), citrate plasma (150 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 to 1000 pg/mL (Cell Culture Supernates), 31.20 to 2000 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Assay Sensitivity 4 pg/mL (Cell Culture Supernates), 11 pg/mL (Serum, Heparin Plasma, Citrate Plasma, EDTA Plasma)
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity Natural and Recombinant Human MIP-1 beta
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 6351
Quantity 1 Plate
Sample Volume Cell Culture Supernates (10uL),Serum (150uL),EDTA Plasma (150uL),Heparin Plasma (150uL),Citrate Plasma (150uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1 b), Small-inducible cytokine A4, T-cell activation protein 2
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.