missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CBLL1 Partial ORF (NP_079090, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_079090 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79872 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184583
|
Abnova™
H00079872-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16194583
|
Abnova™
H00079872-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Epithelial cell cadherin (CDH1; MIM 192090) is endocytosed as a consequence of tyrosine phosphorylation and ubiquitination. HAKAI is an E3 ubiquitin ligase (see UBE3A; MIM 601623) that mediates ubiquitination of the CDH1 complex.[supplied by OMIM]
Sequence: MDHTDNELQGTNSSGSLGGLDVRRRIPIKLISKQANKAKPAPRTQRTINRMPAKAPPGDEEGFDYNEEERYDCKGGELFANQRRFPGHLFWDFQINILGESpecifications
NP_079090 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23109/HAKAI/MGC163401/MGC163403/RNF188 | |
CBLL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79872 | |
CBLL1 (Human) Recombinant Protein (Q01) | |
MDHTDNELQGTNSSGSLGGLDVRRRIPIKLISKQANKAKPAPRTQRTINRMPAKAPPGDEEGFDYNEEERYDCKGGELFANQRRFPGHLFWDFQINILGE | |
RUO | |
CBLL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |