missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CA10 Partial ORF (AAH20577.1, 1 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00056934-Q02.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16055595

  • 508.00 € / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein that belongs to the carbonic anhydrase family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes. The protein encoded by this gene is an acatalytic member of the alpha-carbonic anhydrase subgroup, and it is thought to play a role in the central nervous system, especially in brain development. Multiple transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]

Sequence: MEIVWEVLFLLQANFIVCISAQQNSPKIHEGWWAYKEVVQGSFVPVPSFWGLVNSAWNLCSVGKRQSPVNIETSHMIFDPFLTPLRINTGGRKVSGTMYNTGRHVSLRLDKEHLVNISGGPMTYSHRLEEIRLHFGSEDSQGSEHLLNGQAFSGEVQLIHYNHELYTNVTEAAKSPNGLVVVSIFIKVSDSSNPFLNRMLNRDTITRITYKNDAYLLQGLNIEELYPETSSFITYDGSMTIPPCYETASWIIMNKPVYITRMQMHSLRLLSQNQPSQIF
Specifications

Specifications

AAH20577.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
56.32kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MEIVWEVLFLLQANFIVCISAQQNSPKIHEGWWAYKEVVQGSFVPVPSFWGLVNSAWNLCSVGKRQSPVNIETSHMIFDPFLTPLRINTGGRKVSGTMYNTGRHVSLRLDKEHLVNISGGPMTYSHRLEEIRLHFGSEDSQGSEHLLNGQAFSGEVQLIHYNHELYTNVTEAAKSPNGLVVVSIFIKVSDSSNPFLNRMLNRDTITRITYKNDAYLLQGLNIEELYPETSSFITYDGSMTIPPCYETASWIIMNKPVYITRMQMHSLRLLSQNQPSQIF
RUO
CA10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
56934
CA10 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CA-RPX/CARPX/HUCEP-15
CA10
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions