missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human C1orf83 Full-length ORF (NP_694580.1, 1 a.a. - 208 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00127428-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16137773

  • 508.00 € / 25µg
Estimated Shipment: 04-09-2024
to see stock.

Description

Description

Sequence: MDKFVIRTPRIQNSPQKKDSGGKVYKQATIESLKRVVVVEDIKRWKTMLELPDQTKENLVEALQELKKKIPSREVLKSTRIGHTVNKMRKHSDSEVASLAREVYTEWKTFTEKHSNRPSIEVRSDPKTESLRKNAQKLLSEALELKMDHLLVENIERETFHLCSRLINGPYRRTVRALVFTLKHRAEIRAQVKSGSLPVGTFVQTHKK
Specifications

Specifications

NP_694580.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32112/FLJ39169/RP4-758J24.3
C1orf83
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
127428
C1orf83 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDKFVIRTPRIQNSPQKKDSGGKVYKQATIESLKRVVVVEDIKRWKTMLELPDQTKENLVEALQELKKKIPSREVLKSTRIGHTVNKMRKHSDSEVASLAREVYTEWKTFTEKHSNRPSIEVRSDPKTESLRKNAQKLLSEALELKMDHLLVENIERETFHLCSRLINGPYRRTVRALVFTLKHRAEIRAQVKSGSLPVGTFVQTHKK
RUO
C1orf83
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions