Learn More
Human C1orf43 (NICE3) knockout HEK-293T cell lysate, 1 Kit
Product Code. 32150238 Shop All Abcam Products
Product Code. 32150238 Supplier Abcam Supplier No. ab2587991Kit
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human C1orf43 (NICE3) knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: C1orf43 (NICE3) knockout HEK-293T