missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Bub1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3785-SP
This item is not returnable.
View return policy
Description
Bub1 Polyclonal specifically detects Bub1 in Human samples. It is validated for Western Blot.
Specifications
| Bub1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BUB1 budding uninhibited by benzimidazoles 1 homolog, BUB1Amitotic checkpoint serine/threonine-protein kinase BUB1, BUB1L, budding uninhibited by benzimidazoles 1 (yeast homolog), budding uninhibited by benzimidazoles 1 homolog (yeast), EC 2.7.11.1, hBUB1mitotic spindle checkpoint kinase, putative serine/threonine-protein kinase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 699 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O43683 | |
| BUB1 | |
| E. coli-derived recombinant human Bub1 Met1-Ser250 Accession # O43683 | |
| 25 μg | |
| Primary | |
| Detects human Bub1 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction